missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA12A Recombinant Rabbit Monoclonal Antibody (23GB6030), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552974
This item is not returnable.
View return policy
Description
HSPA12A is a protein coding gene.
Specifications
| HSPA12A | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43301, Q8K0U4 | |
| HSPA12A | |
| A synthesized peptide derived from human HSPA12A (104-143AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB6030 | |
| Unconjugated | |
| HSPA12A | |
| 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 259217, 73442 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction