missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB120-2928-0.025mg
This item is not returnable.
View return policy
Description
HSP90 alpha Polyclonal specifically detects HSP90 alpha in Human, Mouse, Rat, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| HSP90 alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects Heat Shock Protein 86 (HSP 86). This does not detect HSP 84. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 5 μg/mL | |
| P07901 | |
| HSP90AA1 | |
| Synthetic peptide corresponding to residues P(2) E E T Q T Q D Q P M(12) of mouse HSP86. | |
| 0.025 mg | |
| Chaperone Mediated Autophagy (CMA) | |
| 3320 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction