missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
£159.00
Specifications
| Antigen | HSP90 alpha |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HSP90 alpha Polyclonal specifically detects HSP90 alpha in Human, Mouse, Rat, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| HSP90 alpha | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Chaperone Mediated Autophagy (CMA) | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| HSP90AA1 | |
| IgG | |
| Affinity Purified | |
| Detects Heat Shock Protein 86 (HSP 86). This does not detect HSP 84. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P07901 | |
| 3320 | |
| Synthetic peptide corresponding to residues P(2) E E T Q T Q D Q P M(12) of mouse HSP86. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title