missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp70 interacting protein HIP Antibody (CL3708), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61615-25ul
This item is not returnable.
View return policy
Description
Hsp70 interacting protein HIP Monoclonal specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Hsp70 interacting protein HIP | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| ST13 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EITEEMMDQANDKKVAAIEALNDGELQKAIDLFTDAIKLNPRLAILYAKRASVFVKLQKPNAAIRDCDRA | |
| 25 μL | |
| Membrane Trafficking and Chaperones | |
| 6767 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| CL3708 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:10000 - 1:20000 | |
| AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction