missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40/DNAJB1 Antibody (2A7.H6), Novus Biologicals™
Mouse Monoclonal Antibody
£168.00
Specifications
| Antigen | HSP40/DNAJB1 |
|---|---|
| Clone | 2A7.H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot |
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Yeast samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| HSP40/DNAJB1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Yeast | |
| DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
| DNAJB1 | |
| IgG1 κ | |
| Protein G purified | |
| Detects approx 40kDa. Yeast specific. |
| 2A7.H6 | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P25491 | |
| 3337 | |
| Full length protein HSP40 (YDJ1) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title