missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40/DNAJB1 Antibody (2A7.H6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22416-0.025mg
This item is not returnable.
View return policy
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Yeast samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| HSP40/DNAJB1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3337 | |
| Yeast | |
| Purified |
| Western Blot | |
| 2A7.H6 | |
| Unconjugated | |
| P25491 | |
| DNAJB1 | |
| Full length protein HSP40 (YDJ1) | |
| 0.025 mg | |
| Primary | |
| Detects approx 40kDa. Yeast specific. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction