missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £345.00
Specifications
| Antigen | HSP27 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 - 3 ug/mL, Immunohistochemistry-Paraffin 10 ug/mL, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18128701
|
Novus Biologicals
NBP2-24533SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149531
|
Novus Biologicals
NBP2-24533 |
0.1 mg |
£345.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| HSP27 | |
| Western Blot 2 - 3 ug/mL, Immunohistochemistry-Paraffin 10 ug/mL, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| PBS, 0.05% BSA with 0.05% Sodium Azide | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| HSPB1 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Canine, Equine, Primate | |
| P04792 | |
| 3315 | |
| A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title