missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24533SS
This item is not returnable.
View return policy
Description
HSP27 Polyclonal specifically detects HSP27 in Human, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| HSP27 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.05% BSA with 0.05% Sodium Azide | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| 0.5 mg/mL | |
| Western Blot 2 - 3 ug/mL, Immunohistochemistry-Paraffin 10 ug/mL, Knockout Validated | |
| P04792 | |
| HSPB1 | |
| A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody. | |
| 0.025 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 3315 | |
| Human, Canine, Equine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction