missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (967050), Novus Biologicals™
Mouse Monoclonal Antibody
£158.00 - £379.00
Specifications
| Antigen | HSP27 |
|---|---|
| Clone | 967050 |
| Dilution | Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017733
|
Novus Biologicals
MAB1580-SP |
25 μg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665104
|
Novus Biologicals
MAB1580 |
100 μg |
£379.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
| HSP27 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| HSPB1 | |
| IgG1 | |
| Protein A or G purified |
| 967050 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| PBS with No Preservative | |
| 3315 | |
| E. coli-derived recombinant human HSP27 Accession # P04792 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title