missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP27 Antibody (967050), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB1580
This item is not returnable.
View return policy
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| HSP27 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| HSPB1 | |
| E. coli-derived recombinant human HSP27 Accession # P04792 | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| 3315 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 967050 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction