missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSFY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80129
This item is not returnable.
View return policy
Description
HSFY1 Polyclonal specifically detects HSFY1 in Human samples. It is validated for Western Blot.
Specifications
| HSFY1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ25453, Heat shock transcription factor 2-like protein, heat shock transcription factor, Y linked 2, heat shock transcription factor, Y-linked, HSF2L, HSF2-like, HSFY, HSFY1 | |
| Rabbit | |
| 45 kDa | |
| 100 μL | |
| Signal Transduction | |
| 86614 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96LI6 | |
| HSFY1 | |
| Synthetic peptide directed towards the N terminal of human HSFY1 (NP_149099). Peptide sequence SWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKI. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Canine: 79%; Rat: 79%; Mouse: 79%; Bovine: 79%. | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction