missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91313
This item is not returnable.
View return policy
Description
HSF5 Polyclonal specifically detects HSF5 in Human samples. It is validated for Western Blot.
Specifications
| HSF5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ40311, heat shock factor protein 5, heat shock transcription factor 5, heat shock transcription factor family member 5, HSF 5, HSTF 5, HSTF5, MGC134827 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 124535 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001073908 | |
| HSF5 | |
| Synthetic peptide directed towards the middle region of human HSF5. Peptide sequence SKPSEDTGLATPARYREHRSNSQQGKSPDLHLLVDVACKQERFPKEEELK. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Equine: 92%; Mouse: 92%; Pig: 92%; Rabbit: 92%; Chicken: 90%; Guinea pig: 85%. | |
| Human, Mouse, Rat, Pig, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction