missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£159.00 - £483.00
Specifications
| Antigen | HSF1 |
|---|---|
| Dilution | Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay |
| Applications | ELISA, Western Blot, Gel Shift, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009949
|
Novus Biologicals
NBP1-97475-0.025MG |
0.025 mg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759283
|
Novus Biologicals
NBP1-97475-0.05mg |
0.05 mg |
£305.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18180890
|
Novus Biologicals
NBP1-97475-0.2MG |
0.2 mg |
£483.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSF1 Polyclonal specifically detects HSF1 in Human, Mouse, Rat, Gerbil, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Gel Super Shift Assays, Enzyme Immunoassay.Specifications
| HSF1 | |
| ELISA, Western Blot, Gel Shift, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Primate, Rat, Rabbit, Human, Mouse, Gerbil | |
| Q00613 | |
| 3297 | |
| Recombinant human HSF1. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| HSF1 | |
| IgG | |
| Protein A purified | |
| Recognizes nonphosphorylated HSF1 at ∼60kDa and phosphorylated HSF1 at ∼80kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title