missing translation for 'onlineSavingsMsg'
Learn More

HSF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

Brand:  Novus Biologicals NBP1-97475-0.025MG

 View more versions of this product

Product Code. 18009949

  • £159.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSF1 Polyclonal specifically detects HSF1 in Human, Mouse, Rat, Gerbil, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Gel Super Shift Assays, Enzyme Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSF1
Polyclonal
Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay
Q00613
HSF1
Recombinant human HSF1.
0.025 mg
Primary
Recognizes nonphosphorylated HSF1 at ∼60kDa and phosphorylated HSF1 at ∼80kDa.
Store at -20C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot, Gel Shift, Immunoprecipitation
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rabbit
Protein A purified
RUO
3297
Primate, Rat, Rabbit, Human, Mouse, Gerbil
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only