missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Antibody (10H4), Novus Biologicals™
Rat Monoclonal Antibody
£176.00 - £427.00
Specifications
| Antigen | HSF1 |
|---|---|
| Clone | 10H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunohistochemistry 1:8000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:8000, Gel Super Shift Assays |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623928
|
Novus Biologicals
NBP2-42205-0.025mg |
0.025 mg |
£176.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650318
|
Novus Biologicals
NBP2-42205 |
0.1 mg |
£427.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Super Shift AssaysSpecifications
| HSF1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Cancer, DNA Repair, Transcription Factors and Regulators | |
| PBS (pH 7.4), 50% Glycerol | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Purified recombinant mouse HSF1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 10H4 | |
| Western Blot 1:1000, ELISA, Immunohistochemistry 1:8000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:8000, Gel Super Shift Assays | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Q00613 | |
| 3297 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title