missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSF1 Antibody (10H4), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-42205-0.025mg
This item is not returnable.
View return policy
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Super Shift Assays
Specifications
| HSF1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunohistochemistry 1:8000, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:8000, Gel Super Shift Assays | |
| Q00613 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 10H4 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Purified recombinant mouse HSF1 protein | |
| 0.025 mg | |
| Cancer, DNA Repair, Transcription Factors and Regulators | |
| 3297 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction