missing translation for 'onlineSavingsMsg'
Learn More

HSD17B2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen HSD17B2
Concentration 0.29 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16847723
View Documents
Proteintech
10978-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16837723
View Documents
Proteintech
10978-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone.HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
Specifications

Specifications

HSD17B2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
P37059
3294
HSD17B2 Fusion Protein Ag1411
Primary
-20°C
HSD17B2
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
17 beta HSD 2, 17betaHSD2, 20 alpha HSD, E2DH, EDH17B2, HSD17, HSD17B2, SDR9C2
HSD17B2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.