missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10978-1-AP-20UL
This item is not returnable.
View return policy
Description
HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone.HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.Specifications
| HSD17B2 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B2 | |
| 17 beta HSD 2, 17betaHSD2, 20 alpha HSD, E2DH, EDH17B2, HSD17, HSD17B2, SDR9C2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3294 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37059 | |
| HSD17B2 | |
| HSD17B2 Fusion Protein Ag1411 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction