missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsc70 Mouse anti-Human, Mouse, Rat, Clone: 1A4B3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Hsc70 |
|---|---|
| Clone | 1A4B3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16838325
|
Proteintech
66442-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16828325
|
Proteintech
66442-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date.Specifications
| Hsc70 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Heat shock 70 kDa protein 8, heat shock 70kDa protein 8, HSC54, Hsc70, HSC71, HSP71, HSP73, HSPA10, HSPA8, LAP1, NIP71 | |
| HSPA8 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A4B3 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P11142, P63017, P63018 | |
| 15481, 24468, 3312 | |
| Hsc70 Fusion Protein Ag17910 | |
| Primary | |
| -20°C | |
| HSPA8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title