missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsc70 Mouse anti-Human, Mouse, Rat, Clone: 1A4B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66442-1-IG-20UL
This item is not returnable.
View return policy
Description
The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date.Specifications
| Hsc70 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11142, P63017, P63018 | |
| HSPA8 | |
| Hsc70 Fusion Protein Ag17910 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1A4B3 | |
| Unconjugated | |
| HSPA8 | |
| Heat shock 70 kDa protein 8, heat shock 70kDa protein 8, HSC54, Hsc70, HSC71, HSP71, HSP73, HSPA10, HSPA8, LAP1, NIP71 | |
| Mouse | |
| Protein G | |
| RUO | |
| 15481, 24468, 3312 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction