missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
£195.00 - £373.00
Specifications
| Antigen | HRD1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature (PMID 35350536), Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 24453213), Knockout Validated reported in scientific literature (PMID 26551274) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18012044
|
Novus Biologicals
NB100-2526SS |
0.025 mL |
£195.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788193
|
Novus Biologicals
NB100-2526 |
0.1 mL |
£373.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HRD1 Polyclonal specifically detects HRD1 in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| HRD1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Ubiquitin Proteasome Pathway | |
| Whole antisera with No Preservative | |
| E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin | |
| SYVN1 | |
| IgG | |
| Unpurified |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature (PMID 35350536), Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 24453213), Knockout Validated reported in scientific literature (PMID 26551274) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Hamster | |
| Q86TM6 | |
| 84447 | |
| A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title