missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB100-2526SS
This item is not returnable.
View return policy
Description
HRD1 Polyclonal specifically detects HRD1 in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| HRD1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation reported in scientific literature (PMID 35350536), Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 24453213), Knockout Validated reported in scientific literature (PMID 26551274) | |
| Q86TM6 | |
| SYVN1 | |
| A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3] | |
| 0.025 mL | |
| Apoptosis, Cancer, Cell Biology, Ubiquitin Proteasome Pathway | |
| 84447 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| E3 ubiquitin-protein ligase synoviolin, EC 6.3.2, HRD1EC 6.3.2.-, KIAA1810Synovial apoptosis inhibitor 1, MGC40372, synovial apoptosis inhibitor 1, synoviolin | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Hamster | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction