missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR6B/UBE2B Mouse anti-Human, Clone: PCRP-UBE2B-1C7, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | HR6B/UBE2B |
|---|---|
| Clone | PCRP-UBE2B-1C7 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Beskrivning
HR6B/UBE2B Monoclonal specifically detects HR6B/UBE2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifikationer
| HR6B/UBE2B | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins | |
| 10 mM PBS with 0.05% BSA | |
| 7320 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-UBE2B-1C7 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
| Recombinant full-length human HR6B/UBE2B protein | |
| Primary | |
| Store at 4°C. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel