missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR6B/UBE2B Mouse anti-Human, Clone: PCRP-UBE2B-1C7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07169-100ug
This item is not returnable.
View return policy
Description
HR6B/UBE2B Monoclonal specifically detects HR6B/UBE2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| HR6B/UBE2B | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| E2 protein, E2-17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6BEC 6.3.2.19, UBC2, Ubiquitin carrier protein B, ubiquitin-conjugating enzyme E2 B, Ubiquitin-conjugating enzyme E2-17 kDa, ubiquitin-conjugating enzyme E2B (RAD6 homolog), Ubiquitin-protein ligase B | |
| Recombinant full-length human HR6B/UBE2B protein | |
| 100 μg | |
| Biologically Active Proteins | |
| 7320 | |
| Store at 4°C. | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| PCRP-UBE2B-1C7 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction