missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR23A/Rad23A TUBE1 Antibody (SR1997), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | HR23A/Rad23A TUBE1 |
|---|---|
| Clone | SR1997 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651615
|
Novus Biologicals
NBP3-22203-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632494
|
Novus Biologicals
NBP3-22203-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HR23A/Rad23A TUBE1 Monoclonal antibody specifically detects HR23A/Rad23A TUBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| HR23A/Rad23A TUBE1 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 5886 | |
| IgG | |
| Affinity purified |
| SR1997 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| hHR23A, HR23A | |
| A synthesized peptide derived from human HR23A/Rad23A TUBE1 (Uniprot #: P54725) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title