missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HR23A/Rad23A TUBE1 Antibody (SR1997), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22203-100ul
This item is not returnable.
View return policy
Description
HR23A/Rad23A TUBE1 Monoclonal antibody specifically detects HR23A/Rad23A TUBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry
Specifications
| HR23A/Rad23A TUBE1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5886 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry | |
| SR1997 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50 | |
| hHR23A, HR23A | |
| A synthesized peptide derived from human HR23A/Rad23A TUBE1 (Uniprot #: P54725) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction