missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (C1N1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £378.00
Specifications
| Antigen | HPV18 E6 |
|---|---|
| Clone | C1N1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245240
|
Novus Biologicals
NBP2-54807 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657206
|
Novus Biologicals
NBP2-54807-0.025mg |
0.025 mg |
£159.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| HPV18 E6 | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| HPV18E6-beta-galactosidase fusion protein synthesized in E. coli | |
| Primary | |
| 16.5 kDa |
| C1N1 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| IgG | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title