missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (C1N1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54807-0.025mg
This item is not returnable.
View return policy
Description
HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| HPV18 E6 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| Mouse | |
| 16.5 kDa | |
| 0.025 mg | |
| Cancer, Tumor Suppressors | |
| 2475 | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| C1N1 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MTOR | |
| HPV18E6-beta-galactosidase fusion protein synthesized in E. coli | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction