missing translation for 'onlineSavingsMsg'
Learn More

HPV18 E6 Antibody (BF7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£169.00 - £395.00

Specifications

Antigen HPV18 E6
Clone BF7
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000
Applications Western Blot, ELISA, Radioimmune Assays (RIA)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18536231
View Documents
Novus Biologicals
NB100-2769
0.1 mg
£395.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18517931
View Documents
Novus Biologicals
NB100-2769-0.025mg
0.025 mg
£169.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Radioimmune Assays
Specifications

Specifications

HPV18 E6
1 mg/mL
Western Blot, ELISA, Radioimmune Assays (RIA)
Unconjugated
Mouse
Cancer, Tumor Suppressors
PBS
Gel-purified HPV18 E6-beta galactosidase fusion protein.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
BF7
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000
Monoclonal
Purified
RUO
Virus
E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.