missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (BF7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2769-0.025mg
This item is not returnable.
View return policy
Description
HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Radioimmune Assays
Specifications
| HPV18 E6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000 | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| Gel-purified HPV18 E6-beta galactosidase fusion protein. | |
| 0.025 mg | |
| Cancer, Tumor Suppressors | |
| Virus | |
| Purified |
| Western Blot, ELISA, Radioimmune Assays (RIA) | |
| BF7 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction