missing translation for 'onlineSavingsMsg'
Learn More

HPV16 Mouse anti-Virus, Clone: SPM257, Novus Biologicals™

Mouse Monoclonal Antibody

£377.00

Specifications

Antigen HPV16
Clone SPM257
Concentration 0.2 mg/mL
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18687388
View Documents
Novus Biologicals
NBP3-07791-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HPV16
0.2 mg/mL
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Virus
Human Papilloma Virus Type 16
IgG2a κ
Protein A or G purified
SPM257
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Human HPV16, major capsid protein L1
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.