missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 Mouse anti-Virus, Clone: SPM257, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07791-100ug
This item is not returnable.
View return policy
Description
HPV16 Monoclonal specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HPV16 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Human Papilloma Virus Type 16 | |
| Human HPV16, major capsid protein L1 | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgG2a κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM257 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction