missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPa3 Antibody (HIC3-2D12), Novus Biologicals™
Mouse Monoclonal Antibody
£167.00 - £386.00
Specifications
| Antigen | HPa3 |
|---|---|
| Clone | HIC3-2D12 |
| Concentration | 2 mg/ml |
| Dilution | Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308991
|
Bio-Techne
NBP3-18520SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18044845
|
Novus Biologicals
NBP3-18520 |
0.1 mg |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPa3 Monoclonal antibody specifically detects HPa3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| HPa3 | |
| 2 mg/ml | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS | |
| IgM | |
| Ion exchange chromatography |
| HIC3-2D12 | |
| Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human pancreatic islets enriched in non-beta endocine cells. | |
| Primary | |
| Store at 4°CC. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title