missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPa3 Antibody (HIC3-2D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18520
This item is not returnable.
View return policy
Description
HPa3 Monoclonal antibody specifically detects HPa3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HPa3 | |
| Monoclonal | |
| 2 mg/ml | |
| Flow Cytometry, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Human | |
| IgM |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HIC3-2D12 | |
| Unconjugated | |
| PBS | |
| Human pancreatic islets enriched in non-beta endocine cells. | |
| 0.1 mg | |
| Primary | |
| Store at 4°CC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction