missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPa1 Antibody (DHIC2-2C12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£171.00 - £371.00
Specifications
| Antigen | HPa1 |
|---|---|
| Clone | DHIC2-2C12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266550
|
Novus Biologicals
NBP1-18949SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228220
|
Novus Biologicals
NBP1-18949 |
0.1 mL |
£371.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPa1 Monoclonal specifically detects HPa1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| HPa1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| Human pancreatic enriched alpha cells | |
| Primary | |
| Store at 4C. Do not freeze. |
| DHIC2-2C12 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| human pancreatic alpha cell | |
| IgM | |
| Tissue culture supernatant | |
| Specific for alpha endocrine cell types. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title