missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPa1 Antibody (DHIC2-2C12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-18949
This item is not returnable.
View return policy
Description
HPa1 Monoclonal specifically detects HPa1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| HPa1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100 | |
| human pancreatic alpha cell | |
| Human pancreatic enriched alpha cells | |
| 0.1 mL | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| Specific for alpha endocrine cell types. | |
| Store at 4C. Do not freeze. | |
| IgM |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| DHIC2-2C12 | |
| Unconjugated | |
| Tissue culture supernatant with 0.05% Sodium Azide | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction