missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC6 Recombinant Rabbit Monoclonal Antibody (23GB6315), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HOXC6 |
|---|---|
| Clone | 23GB6315 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087818
![]() |
Invitrogen™
MA553020 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087462
|
Invitrogen™
MA553021 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXC6 Recombinant Monoclonal antibody specifically detects HOXC6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HOXC6 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P09630, P10629 | |
| 15425, 252885, 3223 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6315 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Antennapedia-type homeotic factor; CP25; HHO.C8; homeo box 3C; homeo box C6; homeo box C8 protein; homeobox C6; homeobox protein CP25; Homeobox protein HHO.C8; homeobox protein Hox-3.3; Homeobox protein Hox-3C; homeobox protein Hox-6.1; Homeobox protein Hox-C6; Hox C6; HOX3; Hox-3.3; HOX3C; Hox-6.1; Hoxc6; Hoxc-6 | |
| A synthesized peptide derived from human HoxC6 (1-62AA). | |
| Primary | |
| -20°C | |
| HOXC6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title