missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXC6 Recombinant Rabbit Monoclonal Antibody (23GB6315), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553020
This item is not returnable.
View return policy
Description
This protein belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' non-coding exon.
Specifications
| HOXC6 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09630, P10629 | |
| HOXC6 | |
| A synthesized peptide derived from human HoxC6 (1-62AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6315 | |
| Unconjugated | |
| HOXC6 | |
| Antennapedia-type homeotic factor; CP25; HHO.C8; homeo box 3C; homeo box C6; homeo box C8 protein; homeobox C6; homeobox protein CP25; Homeobox protein HHO.C8; homeobox protein Hox-3.3; Homeobox protein Hox-3C; homeobox protein Hox-6.1; Homeobox protein Hox-C6; Hox C6; HOX3; Hox-3.3; HOX3C; Hox-6.1; Hoxc6; Hoxc-6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15425, 252885, 3223 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction