missing translation for 'onlineSavingsMsg'
Learn More

HOXC6 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£161.00 - £383.00

Specifications

Antigen HOXC6
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229457
View Documents
Novus Biologicals
NBP3-33439-100ul
100 μL
£383.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226588
View Documents
Novus Biologicals
NBP3-33439-20ul
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HOXC6 Monoclonal antibody specifically detects HOXC6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

HOXC6
ELISA, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
CP25, HHO.C8, homeo box 3C, homeo box C6, homeo box C8 protein, homeobox C6, Homeobox protein CP25, Homeobox protein HHO.C8, Homeobox protein Hox-3C, homeobox protein Hox-C6, HOX3CHOX3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human HOXC6 (NP_004494.1).,, Sequence:, MNSYFTNPSLSCHLAGGQDVLPNVALNSTAYDPVRHFSTYGAAVAQNRIYSTPFYSPQENVVFSSSRGPYDYGSNSFYQEKDMLSNCRQNTLGHNTQTSI
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol, 0.05% BSA
3223
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.