missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB9 Recombinant Rabbit Monoclonal Antibody (23GB6075), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HOXB9 |
|---|---|
| Clone | 23GB6075 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089585
![]() |
Invitrogen™
MA552980 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089600
|
Invitrogen™
MA552981 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXB9 Recombinant Monoclonal antibody specifically detects HOXB9 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HOXB9 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P17482 | |
| 3219 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6075 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054 | |
| A synthesized peptide derived from human HOXB9 (1-52AA). | |
| Primary | |
| -20°C | |
| HOXB9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title