missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOXB9 Recombinant Rabbit Monoclonal Antibody (23GB6075), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552981
This item is not returnable.
View return policy
Description
HOXB9 belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. HOXB9 is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.
Specifications
| HOXB9 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17482 | |
| HOXB9 | |
| A synthesized peptide derived from human HOXB9 (1-52AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6075 | |
| Unconjugated | |
| HOXB9 | |
| HGNC:5120; homeo box 2E; homeo box B9; homeobox B9; homeobox protein Hox-2.5; Homeobox protein Hox-2E; homeobox protein Hox-B9; HOX2; HOX-2.5; HOX2E; HOXB9; Hoxb-9; MGC124051; MGC124052; MGC124053; MGC124054 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3219 | |
| -20°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering