missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXB6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | HOXB6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229488
|
Novus Biologicals
NBP3-35324-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230808
|
Novus Biologicals
NBP3-35324-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOXB6 Polyclonal antibody specifically detects HOXB6 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| HOXB6 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| homeo box 2B, homeo box B6, homeobox B6, Homeobox protein Hox-2.2, Homeobox protein Hox-2B, homeobox protein Hox-B6, Homeobox protein Hu-2, HOX2, Hox-2.2, HOX2BHU-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-110 of human HOXB6 (NP_061825.2).,, Sequence:, SGYADPLRHYPAPYGPGPGQDKGFATSSYYPPAGGGYGRAAPCDYGPAPAFYREKESACALSGADEQPPFHPEPRKSDCAQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 3216 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title