Learn More
DWK Life Sciences DURAN™ Screw Cap Open Top for Tube Connection, Blue, GL 14, Glass Re-enforced, PP (Screw Connection Cap GL 45 Accessory)
Product Code. 10389462 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 14
Unit Size:
Pack of 2
Product Code. Thread Size (Metric) unitSize
10389462 GL 14 Pack of 2
1 options
Product Code. 10389462 Supplier DWK Life Sciences Supplier No. 1129814
Only null left
Add to basket
Add to basket
Accessory for 2 or 3 port GL 45 connection cap system
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
  • Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
  • Available with 2, or 3 threaded outlets (ports)
  • Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
  • By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Fully compatible with all the GLS 80 threads and therefore highly versatile
Suitable Accessories:
  • 1129750
  • 1129751
  • 1129815
  • 1129816
  • 1129817
  • 1129818
TRUSTED_SUSTAINABILITY
Specifications
Description Screw cap GL 14, PP, for tubing connection
For Use With (Application) Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply
Material Polypropylene, PTFE, Silicone
Quantity 2/Pk.
Thread Size (Metric) GL 14
Neck Finish 12 mm
Autoclavable Autoclavable