missing translation for 'onlineSavingsMsg'
Learn More

Isofield™ Horizon Cleanroom Coverall with Mandarin Collar - ISO Class 4/5 Compatible

Product Code. 32016139
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
32016139 Small
32016140 Medium
32016141 Large
32016142 X-Large
32016143 2X-Large
32016144 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 32016139 Supplier Isofield™ Supplier No. 80820S
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket

Provides essential protection for cleanroom environments with anti-static and low-linting performance.

  • Non-sterile cleanroom coverall with mandarin collar
  • Two-layer mandarin collar provides secure fit at neck area
  • Zip flap with double-sided tape prevents ingress and egress of particles
  • Low-linting, anti-static fabric suitable for chemo handling
  • Elasticated thumb loops
  • Back elastic to prevent bellows effect
  • Aseptically folded
  • PPE Cat III, Type 5/6 certified
  • Manufactured in a facility operating under ISO 9001:2015 quality management system
  • Processed in a NEBB certified ISO Class 5 cleanroom
  • Five-year shelf life from date of manufacture

Technical and Documentation Compliance:

  • COA, COC Available for download online
  • QR Code on cartons allows for immediate document retrieval
  • Helmke Drum Class 1 for cleanliness
  • Static-dissipative
  • Tested against chemotherapy to ASTM F739-20

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Covered Elastic
Neck Style Collared
Hood Type None
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals, Anti-static, Liquid, Particle
Collar Type Mandarin
Back Style Elastic
Latex Free Yes
Model Horizon
No. of Pockets None
Includes Horizon Cleanroom Coverall with Mandarin Collar
Thickness 2.6 mil
For Use With (Application) GMP Cleanroom, Pharmaceutical, Biotechnology, Medical Device, Laboratory, and Electronics Manufacturing Environments
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.