Learn More
Medicom™ Hopen™ Simple White Coveralls
Medicom™ Hopen™ Simple White Coveralls available in a variety of sizes, and styles to fit every need
Brand:  Medicom™ 40.001/03
 View more versions of this product
Product Code. 16564011
  • ONLINE EXCLUSIVE PRICE
    £59.00
    £43.36 / Pack of 25
    Save £15.64
    26% Off
Estimated Shipment: 26-08-2026
Add to basket
Description
Description
  • Single use, white coveralls can be used for a wide variety of industrial needs
  • Individually packaged for hygiene, and controlled distribution purposes
  • Suitable to HACCP procedures
Specifications
Specifications
Large
Polypropylene
Yes
Non-sterile
None
Unisex
White
Zipper
Manufactured under an ISO 9001 Certified Quality System
40 g/m2