missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOP Antibody (S06-4F8), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | HOP |
|---|---|
| Clone | S06-4F8 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643454
|
Novus Biologicals
NBP3-19962-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655154
|
Novus Biologicals
NBP3-19962-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOP Monoclonal antibody specifically detects HOP in Mouse, Rat samples. It is validated for Western BlotSpecifications
| HOP | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO | |
| Recombinant protein of mouse HOP (Uniprot # Q8R1H0) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-4F8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 84525 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title