missing translation for 'onlineSavingsMsg'
Learn More

HOP Antibody (S06-4F8), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-19962-50ul

 View more versions of this product

Product Code. 18643454

  • £328.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HOP Monoclonal antibody specifically detects HOP in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HOP
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Purified
Western Blot
S06-4F8
Unconjugated
CAMEO, HOD, homeodomain-only protein, HOP homeobox, HOPLAGYNECC1OB1SMAP31, Lung cancer-associated Y protein, MGC20820, not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, odd homeobox 1 protein, Odd homeobox protein 1, TOTO
Recombinant protein of mouse HOP (Uniprot # Q8R1H0)
50 μg
Cancer
84525
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.