Learn More
Honeywell™ Supervizor Face Shields
Robust, heavy-duty, helmet-mounted face shield range with hearing protection compatibility. Sperian™ Honeywell™ Supervizor Face Shields have a wide visor selection that can be tailored to the specific hazard/application.
Brand: Honeywell™ 1002297+1002312
Code : S1
Additional Details : Weight : 0.20000kg
Description
- Recommended for use in the chemical and forestry industries, among others
- Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
- Maximum protection, allowing full unrestricted head movement
- Creased visor eliminates gaps and visor profile follows contours of the face
Specifications
EN 166 1B | |
Supervizor Faceshield | |
1 Ea. |
Browguard with Ratchet Headband and 230mm Clear Polycarbonate Visor | |
SB600 and SV9PC | |
Polycarbonate |