missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Spacel™ 3000 RA/EBJ Coveralls with Hood

Product Code. 15219968
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 15219968

Brand: Honeywell™ 4503000L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Spacel™ 3000 RA/EBJ Coveralls are made from three-layer polyethylene film with good abrasion, tear, and perforation resistance. With an elasticated hood, wrists, and ankles plus sealed seams and a sealed zipper/adhesive flap closure, the liquid-tight design protects workers from chemical hazards.

  • Product Type: Coveralls
  • Material: Polyethylene
  • Gender: Unisex
  • Color: Yellow
  • Disposable: Yes
  • Sterility: Non-Sterile
  • Resistance: Chemical, Abrasion, Tear, and Perforation
  • Certification: EC Category 3, Protection Level: 3, 4, 5, 6b
  • Hood, Wrist, and Ankle Type: Elasticated
  • Closure Type: Sealed Zipper/Double Adhesive Flap
  • Seam Type: Sealed
  • For Use With: Chemistry, Petrochemistry, In-Shore/Off-Shore Oil Industry, Industrial Maintenance, Pharmaceutical Industry, Spoiled Beach Cleaning, Transportation of Hazardous Materials, Industrial Sub-Contracting Cleaning, Sanitation, Waste Processing, Emergency Services
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Large
Gender Unisex
Material Polyethylene
Color Yellow
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Elastic
Closure Type Sealed Zipper with Double-adhesive Flap
Seam Type Sealed
Sterility Non-sterile
Protection Class Protection Level: 3, 4, 5, 6b
Resistance Chemicals, Abrasion, Tears, Perforation
Construction Three-Layer Film
Coating Polyethylene Film
Certifications/Compliance (Europe) EC Category: 3
Thickness 100 μm
For Use With (Application) Chemistry, Petrochemistry, In-Shore/Off-Shore Oil Industry, Industrial Maintenance, Pharmaceutical Industry, Spoiled Beach Cleaning, Transportation of Hazardous Materials, Industrial Sub-Contracting Cleaning, Sanitation, Waste Processing, Emergency Services
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.