Learn More
Sperian™ Honeywell™ LG Safety Goggles
Product Code. 12980385
Change view
Click to view available options
:
Clear Uncoated Polycarbonate Lens
Product Code.
12980385 Clear Uncoated Polycarbonate Lens
1 options
Product Code. 12980385 Supplier Sperian™ Supplier No. 1005507
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An impact-resistant polycarbonate lens pair of goggles for excellent eye protection. Sperian™ Honeywell™ LG Safety Goggles are an excellent barrier against flying particles, coarse dusts, liquid droplets, mists and sprays.
  • Recommended for manufacturing and pharmaceutical work
  • Lenses are colorless and anti-fog
  • With 4 indirect vents and a wide field of vision
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Spectacles
For Use With (Equipment) Industry Automotive and Part Manufacturer Building and Construction Food Industries Industrial Cleaning Laboratory Maintenance
For Use With (Application) Industry Automotive and Part Manufacturer Building and Construction Food Industries Industrial Cleaning Laboratory Maintenance
Frame Color Clear
No. per Pack 1 Ea.
Material Polycarbonate